Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Cuvettes
Spectrophotometer Cuvettes
VWR®, Square Cuvettes
VWR®, Square Cuvettes
Cuvette, square cuvettes, glass, path length 30 mm, 2 pcs
634-6015
:
VWR Collection
VWR®, Square Cuvettes
634-6015
:
VWR Collection
VWR®, Square Cuvettes
Cuvette, square cuvettes, glass, path length 30 mm, 2 pcs
102,00 €
1 * 2 items
-
+
CoC VWR® V-1200 UV-1600PC UV-3100PC
Manual P6 Spectrophotometer
VWR-UV-3100PC-Manual-V1_EN_060910.pdf
VWR® UV-1600PC
VWR® UV-6300PC_260515
VWR® V-1200
TDS VWR Glass Cuvette,10 mm.pdf
TDS VWR Glass Cuvette,100 mm.pdf
TDS VWR Glass Cuvette,20 mm.pdf
TDS VWR Glass Cuvette,30 mm.pdf
TDS VWR Glass Cuvette,50 mm.pdf
VWR Products for Nucleic Acid Preparation
Description:
Cuvette, square cuvettes, glass, path length 30 mm, 2 pcs
Seq:
3