Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
PH ELECTRODE 1M CABLE 1 * 1 items
PH ELECTRODE 1M CABLE 1 * 1 items
Print
PH ELECTRODE 1M CABLE 1 * 1 items
Catalog # CONSSP20T
Supplier:
CONSORT
PH ELECTRODE 1M CABLE 1 * 1 items
Catalog # CONSSP20T
Supplier:
CONSORT
Expand image
Return to Top
PH ELECTRODE 1M CABLE 1 * 1 items
Add to cart
Price
147,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents