Pipette tips, ratiolab®
Type ES, 500 - 2500 µl, Transparent, Eppendorf® Reference®, Socorex® Calibra, 250/bag
Catalog # RATI2500180
Supplier: RATIOLAB
Jump to:
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifications
- Packed:250/bag
- Volume:500 - 2500 µl
- Seq:52
- Tip style:Type ES
- Tip colour:Transparent
- For use with:Eppendorf® Reference®, Socorex® Calibra