Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Absorbent Bench Underpads

VWR® Absorbent Bench Underpads

Deluxe bench underpads, Blue, 432×610 mm, 340 ml
Catalog # VWRU56616-018
Supplier:   VWR Collection
VWR® Absorbent Bench Underpads
Catalog # VWRU56616-018
Supplier:  VWR Collection
VWR® Absorbent Bench Underpads
VWR® Absorbent Bench Underpads

Deluxe bench underpads, Blue, 432×610 mm, 340 ml

Price
273,00 €
Log in for availability
1 * 300 items
Product Details & Documents

Absorbent underpads with a three-layer construction made from PP top and back sheets and a cellulose filler.

  • Top sheet made from white spunbound polypropylene
  • Absorbent cellulose fluff filler between two layers of tissue for superior wicking of fluid
  • Cushion effect to protect labware from breakage
  • Embossed PP backing sheet comes in different colours
  • Sealed on four sides to prevent liquid seepage

Deluxe pads are more absorbent and thicker with more cellulose fluff than standard pads.

Ordering information: Super Deluxe pads have an added polymer that absorbs more moisture.

Specifications
  • Colour:
    Blue
  • Description:
    Deluxe bench underpads
  • L×W:
    432×610 mm
  • Seq:
    06
  • Absorbency:
    340 ml