Orders LinkContactUsLinkComponent
Nitrile gloves, Omega™ Pro-Tech
Nitrile gloves, Omega™ Pro-Tech
XS, Purple, 240 mm, 10 boxes, 100 each
Catalog # NITROPT2101-XS
Supplier:   Nitritex
Nitrile gloves, Omega™ Pro-Tech
Catalog # NITROPT2101-XS
Supplier:  Nitritex
Nitrile gloves, Omega™ Pro-Tech
Nitrile gloves, Omega™ Pro-Tech
XS, Purple, 240 mm, 10 boxes, 100 each
Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents
These nitrile gloves feature an ambidextrous shape and beaded cuffs. They are resistant to a wide range of chemicals.
  • Non sterile, powder-free
  • Contain no natural latex proteins
  • Textured for improved grip
  • Beaded cuff provides easier donning and stability on the arm
  • Each size carries a unique bar code on the inner dispenser box (EAN-13) and the outer carton (ITF-14), which allows automatic data entry into stock control systems
Certifications: EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010; Cat. III; AQL 1,5
Food contact approved
Specifications
  • Glove size:
    XS
  • Packed:
    10 boxes, 100 each
  • Colour:
    Purple
  • Length:
    240 mm
  • Seq:
    01
  • Cat. No.:
    NITROPT2101-XS
  • Latex / latex-free:
    Latex-free
  • Food contact approved:
    +
  • Disposable / reusable:
    Disposable
  • Model:
    Pro-Tech
  • Cuff style:
    Beaded
  • Powdered / powder-free:
    Powder-free
  • EN norm:
    EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010
  • Finish:
    Textured
  • Ambidextrous / hand-specific:
    Ambidextrous