Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Split, with wool, universal use with rapid autosamplers, for Varian 1075/1077, 6,3 mm, 4,0 mm, 72 mm
Catalog # 568-0275
Supplier:   Restek
Inlet liners
Catalog # 568-0275
Supplier:  Restek
Inlet liners
Inlet liners

Split, with wool, universal use with rapid autosamplers, for Varian 1075/1077, 6,3 mm, 4,0 mm, 72 mm

Price
224,00 €
Log in for availability
1 * 5 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Split, with wool, universal use with rapid autosamplers, for Varian 1075/1077
  • Length:
    72 mm
  • Seq:
    051