Jars, glass
Glass jar with metal locking bracket and rubber seal around the rim, 500 ml
Catalog # SPVAH03064
Supplier: GRAVIS
Jump to:
Product Details & Documents
Soda lime glass jars, suitable for storage of samples in the laboratory.
Specifications
- Capacity:500 ml
- Description:Glass jar with metal locking bracket and rubber seal around the rim
- Seq:01