Orders LinkContactUsLinkComponent
3,3'-Thiodipropionic acid ≥98%
Glass bottle for solids, 1.000 g
  A17220.0B
3,3'-Thiodipropionic acid ≥98%
  A17220.0B
 :  111-17-1
3,3'-Thiodipropionic acid ≥98%
3,3'-Thiodipropionic acid ≥98%
Glass bottle for solids, 1.000 g
 :  111-17-1
60,10 €
1 * 1.000 g
Purity: 98% CAS No.: 111-17-1 Molecular Formula: C6H10O4S Formula Weight: 178.21 Melting Point: 127-134° Beilstein Reference: 1210299 Merck Reference: 14,9332 MDL No.: MFCD00002781
 :  3,3-Thiodipropionic acid
  •  :111-17-1
  •  :C₆H₁₀O₄S
  •  :MFCD00002781
  •  :178.21 g/mol
  •  :14,9332
  •  :347 °C (1013 hPa)
  •  :127…134 °C
  •  :1.496 g/cm³ (20 °C)
  •  :Ambient
  •  :203-841-3
  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g