Orders LinkContactUsLinkComponent
Accessories for TW alpha plus
Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)
Catalog # SCOI285213884
Supplier:   SI Analytics
Accessories for TW alpha plus
Catalog # SCOI285213884
Supplier:  SI Analytics
Accessories for TW alpha plus
Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)
Price
868,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)