Orders LinkContactUsLinkComponent
Pre-prepared polystyrene kits, EasiCal™
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-1, 580 - 7500000 g/mol, 5
Catalog # VARIPL2010-0505
Supplier:   VARIAN
Pre-prepared polystyrene kits, EasiCal™
Catalog # VARIPL2010-0505
Supplier:  VARIAN
CAS Number:  
Pre-prepared polystyrene kits, EasiCal™
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-1, 580 - 7500000 g/mol, 5
CAS Number:  
Price
1 831,17 €
Log in for availability
1 * 5 items
Product Details & Documents
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
  • Easy three step process with no fuss
  • Cost effective
  • Only two injections for improved productivity
  • Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.
Specifications
Specifications
  • Pk:
    5
  • Description:
    Polystyrene PS-1, 580 - 7500000 g/mol
Specifications