Desiccator plates
Porcelain, 300 mm
Catalog # HECH41175030
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Porcelain plates for desiccators with many small holes, without base.
Specifications
- Ø:300 mm
- Type:Porcelain
Porcelain plates for desiccators with many small holes, without base.