Order Entry
Export
Orders LinkContactUsLinkComponent
Serum pipettes

Serum pipettes

Serum pipettes, graduated, 2,5 ml, 153 mm, -, 500/inner pack, 1500 pieces/carton
Catalog # GREI612301_500
Serum pipettes
Catalog # GREI612301_500
Supplier:  
Serum pipettes

Serum pipettes, graduated, 2,5 ml, 153 mm, -, 500/inner pack, 1500 pieces/carton

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents

These pipettes are suitable for the rapid transfer of liquids.

Specifications
  • Packed:
    500/inner pack, 1500 pieces/carton
  • Sterile:
    -
  • Capacity:
    2,5 ml
  • Description:
    Serum pipettes, graduated
  • Length:
    153 mm
  • Seq:
    18