Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dimethylthiophene ≥97%

Glass bottle for solids, 5 g
Catalog # H31641.06
Flammable
Irritant
2,3-Dimethylthiophene ≥97%
Catalog # H31641.06
CAS Number:  632-16-6
2,3-Dimethylthiophene ≥97%
2,3-Dimethylthiophene ≥97%

Glass bottle for solids, 5 g

CAS Number:  632-16-6
Price
363,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 632-16-6 Molecular Formula: C6H8S Formula Weight: 112.19 Flash Point: 26° (79°F) Boiling Point: 142° UN No.: 1993 MDL No.: MFCD00130081

  • CAS Number:632-16-6
  • Formula:C₆H₈S
  • MDL Number:MFCD00130081
  • Molecular Weight:112.2 g/mol
  • Boiling Pt:142 °C
  • Melting Pt:–49 °C
  • Density:1.0021 g/cm³ (20 °C)
  • Flash Pt:26 °C (79 °F)
  • Storage Temperature:Ambient
  • EINECS:211-170-2
  • ADR:3,III
  •  :Density: 1,0021 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications