2,3-Dimethylthiophene ≥97%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
2,3-Dimethylthiophene ≥97%
Catalog # H31641.03
Supplier: Thermo Fisher Scientific
CAS Number: 632-16-6
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 632-16-6 Molecular Formula: C6H8S Formula Weight: 112.19 Flash Point: 26° (79°F) Boiling Point: 142° UN No.: 1993 MDL No.: MFCD00130081
- CAS Number:632-16-6
- Formula:C₆H₈S
- MDL Number:MFCD00130081
- Molecular Weight:112.2 g/mol
- Boiling Pt:142 °C
- Melting Pt:–49 °C
- Density:1.0021 g/cm³ (20 °C)
- Flash Pt:26 °C (79 °F)
- Storage Temperature:Ambient
- EINECS:211-170-2
- ADR:3,III
- :Density: 1,0021 g/cm³ (20 °C)
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



