ContactUsLinkComponent
Pcr tube, 0.2ml, thin wall, clear, domed cap, non-sterile, autoclavable, virgin polypropylene, dnase, rnase, and pyrogen-free 1 * 1.000 items
Catalog # BITX3425.X
Supplier:  BIOTIX
undefined
Pcr tube, 0.2ml, thin wall, clear, domed cap, non-sterile, autoclavable, virgin polypropylene, dnase, rnase, and pyrogen-free 1 * 1.000 items
Catalog # BITX3425.X
Supplier:  BIOTIX