Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Combs for shiroGEL Horizontal Gel Systems

VWR® Combs for shiroGEL Horizontal Gel Systems

Comb, 28-well, 0,75 mm thickness for 15 cm gel tray*
Catalog # 700-0282
Supplier:   VWR Collection
VWR® Combs for shiroGEL Horizontal Gel Systems
Catalog # 700-0282
Supplier:  VWR Collection
VWR® Combs for shiroGEL Horizontal Gel Systems
VWR® Combs for shiroGEL Horizontal Gel Systems

Comb, 28-well, 0,75 mm thickness for 15 cm gel tray*

Price
150,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Documents

Specifications
  • Description:
    Comb, 28-well, 0,75 mm thickness for 15 cm gel tray*
  • Seq:
    25