Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Combs for shiroGEL Horizontal Gel Systems

VWR® Combs for shiroGEL Horizontal Gel Systems

Comb, 10-well, 1,0 mm thickness for 15 cm gel tray
Catalog # 700-0265
Supplier:   VWR Collection
VWR® Combs for shiroGEL Horizontal Gel Systems
Catalog # 700-0265
Supplier:  VWR Collection
VWR® Combs for shiroGEL Horizontal Gel Systems
VWR® Combs for shiroGEL Horizontal Gel Systems

Comb, 10-well, 1,0 mm thickness for 15 cm gel tray

Price
66,90 €
Log in for availability
1 * 1 items
Product Details & Documents

Documents

Specifications
  • Description:
    Comb, 10-well, 1,0 mm thickness for 15 cm gel tray
  • Seq:
    05