Order Entry
Export
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ SAX

Solid phase extraction cartridges, HyperSep™ SAX

Cartridge, SAX, 40 - 60 µm, 6 ml, 500 mg
Catalog # HYPE60108-360
Solid phase extraction cartridges, HyperSep™ SAX
Catalog # HYPE60108-360
Solid phase extraction cartridges, HyperSep™ SAX
Solid phase extraction cartridges, HyperSep™ SAX

Cartridge, SAX, 40 - 60 µm, 6 ml, 500 mg

Price
248,00 €
Log in for availability
1 * 30 items
Product Details & Documents

Strong anion exchange sorbent for extraction of weak acids.

  • Extracts negatively charged compounds from both aqueous and non-aqueous solutions
  • Ideally suited for the extraction of weak acids, e.g. carboxylic acids
  • Amine group masks the effect of the carbon chain of the functional group
  • Selectivity can be tuned by selection of buffer
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    6 ml
  • Seq:
    05
  • Bed weight:
    500 mg
  • Phase:
    SAX