Solid phase extraction cartridges, HyperSep™ SAX
Cartridge, SAX, 40 - 60 µm, 3 ml, 200 mg
Catalog # HYPE60108-419
Supplier: Thermo Fisher Scientific
Solid phase extraction cartridges, HyperSep™ SAX
Catalog # HYPE60108-419
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Strong anion exchange sorbent for extraction of weak acids.
- Extracts negatively charged compounds from both aqueous and non-aqueous solutions
- Ideally suited for the extraction of weak acids, e.g. carboxylic acids
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by selection of buffer
Specifications
- Column Format:Cartridge
- Particle Size:40 - 60 µm
- Capacity:3 ml
- Seq:03
- Bed weight:200 mg
- Phase:SAX



