Order Entry
Export
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 3 ml, 500 mg
  HYPE60108-423
Solid phase extraction cartridges, HyperSep™ SCX
  HYPE60108-423
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 3 ml, 500 mg

 
260,00 €
   
1 * 50 items
 

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
 
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    3 ml
  • Seq:
    04
  • Bed weight:
    500 mg
  • Phase:
    SCX