Order Entry
Export
Orders LinkContactUsLinkComponent
 

pH Standard Solutions, IUPAC, Certified

10,012, ±0,010, 25 °C, 500 mL
Catalog # 665-0398
Supplier:   Hach
pH Standard Solutions, IUPAC, Certified
Catalog # 665-0398
Supplier:  Hach
CAS Number:  
pH Standard Solutions, IUPAC, Certified
pH Standard Solutions, IUPAC, Certified

10,012, ±0,010, 25 °C, 500 mL

CAS Number:  
Price
62,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Buffers traceable to IUPAC/NIST pH scale.

  • For calibration of pH meters
  • Accuracy at 25 °C: ±0,010 or ±0,05 (pH 12,45)

Each standard is delivered in an airtight package ensuring safe storage. The nominal value of the pH standard is guaranteed until first opening, certified value is guaranteed for two years from the date of issue of the certificate.

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,010
  • pH value:
    10,012

Specifications