Orders LinkContactUsLinkComponent
PTFE balls, BOLA
PTFE balls, BOLA
Balls, PTFE, 15 mm
Catalog # BOHLH964-15
Supplier:   Bohlender
PTFE balls, BOLA
Catalog # BOHLH964-15
Supplier:  Bohlender
PTFE balls, BOLA
PTFE balls, BOLA
Balls, PTFE, 15 mm
Price
160,00 €
Log in for availability
1 * 50 items
Product Details & Documents
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
Specifications
  • Ø:
    15 mm
  • Description:
    Balls
  • Material:
    PTFE
  • Seq:
    008