Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Alternate analytes B solution, 5% HNO₃, 100 mg/L: Al, As, B, Mo, Na, Sb, Se, Tl; 10 mg/L: Ca, Fe, Mg, Si, 1
  PERCN9300229
Interference check standards for ICP-OES
  PERCN9300229
 :  
Interference check standards for ICP-OES
Alternate analytes B solution, 5% HNO₃, 100 mg/L: Al, As, B, Mo, Na, Sb, Se, Tl; 10 mg/L: Ca, Fe, Mg, Si, 1
 :  
309,00 €
1 * 1 items
  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    Alternate analytes B solution
  • Contents:
    100 mg/L: Al, As, B, Mo, Na, Sb, Se, Tl; 10 mg/L: Ca, Fe, Mg, Si