Interference check standards for ICP-OES
Alternate interferents A solution, 5% HNO₃, 1,000 mg/L: Cr, Cu, Mn,Ni, Ti, V, 1
Catalog # PERCN9300228
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Matrix:5% HNO₃
- Pk:1
- Description:Alternate interferents A solution
- Contents:1,000 mg/L: Cr, Cu, Mn,Ni, Ti, V