Order Entry
Export
Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES

Interference check standards for ICP-OES

Analytes B solution, 5% HNO₃, 100 mg/L: Cd, Ni, Zn; 60 mg/L: Sb; 50 mg/L: Ba, Be, Co, Cr, Cu, Mn, V; 20 mg/L: Ag; 10 mg/L: As, Tl; 5 mg/L: Pb, Se;, 1
Catalog # PERCN9300227
Supplier:   PerkinElmer
Interference check standards for ICP-OES
Catalog # PERCN9300227
Supplier:  PerkinElmer
CAS Number:  
Interference check standards for ICP-OES

Analytes B solution, 5% HNO₃, 100 mg/L: Cd, Ni, Zn; 60 mg/L: Sb; 50 mg/L: Ba, Be, Co, Cr, Cu, Mn, V; 20 mg/L: Ag; 10 mg/L: As, Tl; 5 mg/L: Pb, Se;, 1

CAS Number:  
Price
296,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    Analytes B solution
  • Contents:
    100 mg/L: Cd, Ni, Zn; 60 mg/L: Sb; 50 mg/L: Ba, Be, Co, Cr, Cu, Mn, V; 20 mg/L: Ag; 10 mg/L: As, Tl; 5 mg/L: Pb, Se;

Specifications