Interference check standards for ICP-OES
Interferents A solution, 5% HNO₃, 5,000 mg/L: Al, Ca, Mg; 2,000 mg/L: Fe, 1
Catalog # PERCN9300226
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Matrix:5% HNO₃
- Pk:1
- Description:Interferents A solution
- Contents:5,000 mg/L: Al, Ca, Mg; 2,000 mg/L: Fe