Interference check standards for ICP-OES
Interference check standard 5, 5% HNO₃, 6,000 mg/L: Ca; 5,000 mg/L: Fe; 3,000 mg/L: Mg; 1,200 mg/L: Al; 1,000 mg/L: Na, 1
Catalog # PERCN9300208
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Matrix:5% HNO₃
- Pk:1
- Description:Interference check standard 5
- Contents:6,000 mg/L: Ca; 5,000 mg/L: Fe; 3,000 mg/L: Mg; 1,200 mg/L: Al; 1,000 mg/L: Na