Interference check standards for ICP-OES
Interference check solution 2, 2% HNO₃, 10 mg/L: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn, 1
Catalog # PERCN9303830
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Matrix:2% HNO₃
- Pk:1
- Description:Interference check solution 2
- Contents:10 mg/L: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn