Interference check standards for ICP-OES
Interference check solution 1, 10,000 mg/L: Cl; 2000 mg/L: C; 1000 mg/L: Al, Ca, Fe, K, Mg, Na, P, S; 20 mg/L: Mo, Ti, 1
Catalog # PERCN9303828
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Interference check solution 1
- Contents:10,000 mg/L: Cl; 2000 mg/L: C; 1000 mg/L: Al, Ca, Fe, K, Mg, Na, P, S; 20 mg/L: Mo, Ti