Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Interference check standards, 1000 mg/L: Sb, 1
Catalog # PERCN9303797
Supplier:   PerkinElmer
Interference check standards for ICP-OES
Catalog # PERCN9303797
Supplier:  PerkinElmer
CAS Number:  
Interference check standards for ICP-OES
Interference check standards, 1000 mg/L: Sb, 1
CAS Number:  
Price
129,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
Specifications
  • Pk:
    1
  • Description:
    Interference check standards
  • Contents:
    1000 mg/L: Sb
Specifications