Graphite contact cylinders
THGA contact cylinders, modified, 5 pairs
Catalog # PERCB3002102
Supplier: PerkinElmer
Jump to:
Product Details & Documents
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
- Made from a proprietary blend of base graphite material
- Produced within tight tolerances to guaranty exact fitment and maximize performance
- Modified contacts used in applications to minimize condensation and improve analytical performance with difficult matrices (e.g., 2% Ni solutions, edible oils, etc.)
The proper performance, maintenance and serviceability of PerkinElmer graphite furnace systems can be assured only through the use of genuine PerkinElmer supplied graphite components such as these contact cylinders.
Fits PerkinElmer PinAAcle 900T, PinAAcle 900Z, AAnalyst 600, AAnalyst 800, Model 4100ZL, Model 4110ZL, Model 5100ZL, SIMAA 6000 Zeeman AA instruments equipped with a graphite furnace.
Specifications
- Packed:5 pairs
- Description:THGA contact cylinders, modified
- Seq:04