Order Entry
Export
Orders LinkContactUsLinkComponent
GET™ CLEAN DNA Kit

GET™ CLEAN DNA Kit

GET™ CLEAN DNA Kit, 50 preps, 1 KIT
Catalog # 786-356
Supplier:   G-Biosciences
GET™ CLEAN DNA Kit
Catalog # 786-356
Supplier:  G-Biosciences
CAS Number:  
GET™ CLEAN DNA Kit
GET™ CLEAN DNA Kit

GET™ CLEAN DNA Kit, 50 preps, 1 KIT

CAS Number:  
Price
123,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

GET™ CLEAN DNA uses high binding affinity GET™ spin columns to remove salts, enzymes, unincorporated nucleotides, radiolabels, and primer-dimers from any DNA preparation of 100 bp to 20 kb. GET™ spin columns has an enhanced binding affinity for DNA, thus eliminating loss or damage of DNA.

  • Spin column format
  • Enhanced DNA binding affinity
  • Protocol takes 5 to 10 minutes
  • No phenol/chloroform extraction or alcohol precipitations
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    GET™ CLEAN DNA Kit, 50 preps
  • Cat. No.:
    786-356
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Description:
    GET™ CLEAN DNA Kit
  • Sample type:
    PCR and other enzymatic reactions

Specifications