0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
MW SIZING STANDARD 1 * 3 items
MW SIZING STANDARD 1 * 3 items
BECLA22196
:
Beckman Coulter
MW SIZING STANDARD 1 * 3 items
BECLA22196
:
Beckman Coulter
MW SIZING STANDARD 1 * 3 items
1 180,00 €
1 * 3 items
-
+