Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methyl oleate 96%

Glass bottle for solids, 100 g
Catalog # H31358.22
Methyl oleate 96%
Catalog # H31358.22
CAS Number:  112-62-9
Methyl oleate 96%
Methyl oleate 96%

Glass bottle for solids, 100 g

CAS Number:  112-62-9
Price
540,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 96% CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578

Synonyms:  cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
  • CAS Number:112-62-9
  • Formula:C₁₉H₃₆O₂
  • MDL Number:MFCD00009578
  • Molecular Weight:296.49 g/mol
  • Merck Index:14,6828
  • Boiling Pt:168…170 °C (2 mmHg)
  • Melting Pt:–20 °C
  • Density:0.876
  • Flash Pt:163 °C (325 °F)
  • Storage Temperature:Ambient
  • EINECS:203-992-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications