Methyl oleate 96%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578
Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
- CAS Number:112-62-9
- Formula:C₁₉H₃₆O₂
- MDL Number:MFCD00009578
- Molecular Weight:296.49 g/mol
- Merck Index:14,6828
- Boiling Pt:168…170 °C (2 mmHg)
- Melting Pt:–20 °C
- Density:0.876
- Flash Pt:163 °C (325 °F)
- Storage Temperature:Ambient
- EINECS:203-992-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



