Single channel pipettes, mechanical, fixed / variable volume, labopette®
500 - 5000 µl, 10 µl, ±0,8 %, <0,3 %, Clear, 100 - 500 µl
Catalog # HIRS9475405
Supplier: HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Catalog # HIRS9475405
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
The reliable labopette® pipette features superior ergonomic characteristics, right down to the smallest detail. The entire working process is optimised to ensure non fatiguing work that places a minimum of stress on the joints.
- Volume configuration with digital locking feature
- With tip ejection and calibration option
- Tip seal and safety filter to prevent contamination
- Trigger grip ensures a firm but relaxed hold during pipetting
- Ergonomic design of the handle offers an equally firm, comfortable hold for large and small hands, right and left-handed users and hands with or without gloves
- Effective thermal insulation of the unit interior prevents hand heat penetrating into the interior of the unit
- Completely autoclavable without dismantling
Delivery information: Supplied with calibration tool, pipette holder, individual serial number and calibration certificate.
Specifications
- Volume:500 - 5000 µl
- Seq:08
- Increment:10 µl
- Accuracy:±0,8 %
- Imprecision:<0,3 %
- Recommended tips:Clear, 100 - 500 µl
Accessories
Product Information
- Catalog #AvailabilityPrice
- Safe-Cone filters, for labopette® variable 1000 - 5000 µl; fix 2000/5000 µlSafe-Cone filters, for labopette® variable 1000 - 5000 µl; fix 2000/5000 µlHIRS9470010Log in for availability1 * 50 items - 58,90 €
Product Information
- Catalog #AvailabilityPrice
- Safe-Cone filters, for labopette® variable 2 - 20/5 - 50 µl; fix 20/25/50 µlSafe-Cone filters, for labopette® variable 2 - 20/5 - 50 µl; fix 20/25/50 µlHIRS9470013Log in for availability1 * 50 items - 58,90 €
Product Information
- Catalog #AvailabilityPrice
- Safe-Cone filters, for labopette® elctronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; variable 100 - 1000 µl; fix 250/500/1000 µlSafe-Cone filters, for labopette® elctronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; variable 100 - 1000 µl; fix 250/500/1000 µlHIRS9470011Log in for availability1 * 50 items - 58,90 €
Product Information
- Catalog #AvailabilityPrice
- Safe-Cone filters, for labopette® variable 10 - 100/20 - 200 µl; fix 100/200 µlSafe-Cone filters, for labopette® variable 10 - 100/20 - 200 µl; fix 100/200 µlHIRS9470012Log in for availability1 * 50 items - 58,90 €
Product Information
- Catalog #AvailabilityPrice
- DKD calibration certificate for labopette® fix 0,5 μl - 5 mlDKD calibration certificate for labopette® fix 0,5 μl - 5 mlHIRS9980116Log in for availability1 * 1 items - 91,00 €
Product Information
- Catalog #AvailabilityPrice
- DKD calibration certificate for labopette® 12-channel, 5 - 250 μlDKD calibration certificate for labopette® 12-channel, 5 - 250 μlHIRS9980118Log in for availability1 * 1 items - 395,00 €
Product Information
- Catalog #AvailabilityPrice
- Stand for six mechanical pipettes
Product Information
- Catalog #AvailabilityPrice
- Carousel stand for six mechanical pipettesCarousel stand for six mechanical pipettesHIRS9470004Log in for availability1 * 1 items - 175,00 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94901010,2 - 10 µlTransparentHirschmann Laborgeräte, Gilson® pipettesLog in for availability1 * 1.000 items - 34,00 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94901021 - 200 µlYellowHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog in for availability1 * 1.000 items - 22,70 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS9490103100 - 1000 µlBlueHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog in for availability1 * 1.000 items - 25,30 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94901061 - 250 µlTransparentlabopette® multichannel pipettesLog in for availability1 * 1.000 items - 36,80 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS9490124100 - 5000 µlTransparentHirschmann Laborgeräte pipettesLog in for availability1 * 300 items - 54,00 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94902010,2 - 10 µlTransparentHirschmann Laborgeräte, Gilson® pipettesLog in for availability1 * 960 items - 81,00 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94902021 - 200 µlYellowHirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettesLog in for availability1 * 960 items - 61,10 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS949020550 - 1200 µlTransparentHirschmann Laborgeräte pipettesLog in for availability1 * 960 items - 151,00 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94902061 - 250 µlTransparentlabopette® multichannel pipettesLog in for availability1 * 960 items - 60,70 €
Product Information
- Catalog #VolumeTip colourFor use withAvailabilityPrice
- HIRS94901041 - 10000 µlTransparentHirschmann Laborgeräte pipettesLog in for availability1 * 100 items - 54,00 €
Product Information
- Catalog #Tip styleFor use withAvailabilityPrice
- HIRS9490109Multibox, autoclavablerefill trays of pipette tips up to 300 µlLog in for availability1 * 1 items - 29,00 €
Product Information
- Catalog #Tip styleFor use withAvailabilityPrice
- HIRS9490110Multibox, autoclavablerefill trays of pipette tips up to 1000 µlLog in for availability1 * 1 items - 30,60 €
Product Information
- Catalog #AvailabilityPrice
- Safe-Cone filters, for labopette® fix 10/20/25/250 µl; multi-channel 5,0 - 50/50 - 300 µl; electronic multi-channel 25 - 250/5 - 100 µlSafe-Cone filters, for labopette® fix 10/20/25/250 µl; multi-channel 5,0 - 50/50 - 300 µl; electronic multi-channel 25 - 250/5 - 100 µlHIRS9470014Log in for availability1 * 50 items - 58,90 €
Product Information
- Catalog #AvailabilityPrice
- DKD calibration certificate for labopette® variable volumes 0,5 μl - 5 ml / labopettorDKD calibration certificate for labopette® variable volumes 0,5 μl - 5 ml / labopettorHIRS9980115Log in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #AvailabilityPrice
- DKD calibration certificate for labopette® 8-channel, 0,5 - 250 μlDKD calibration certificate for labopette® 8-channel, 0,5 - 250 μlHIRS9980117Log in for availability1 * 1 items - 352,00 €