Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Difluoroacetone ≥98%

Glass bottle for solids, 5 g
Catalog # H51145.06
Toxic
Flammable
1,3-Difluoroacetone ≥98%
Catalog # H51145.06
CAS Number:  453-14-5
1,3-Difluoroacetone ≥98%
1,3-Difluoroacetone ≥98%

Glass bottle for solids, 5 g

CAS Number:  453-14-5
Price
404,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 453-14-5 Molecular Formula: C3H4F2O Formula Weight: 94.06 Boiling Point: 101-102° UN No.: 2929 MDL No.: MFCD07368665

  • CAS Number:453-14-5
  • Formula:C₃H₄F₂O
  • MDL Number:MFCD07368665
  • Molecular Weight:94.06 g/mol
  • Boiling Pt:101…102 °C
  • Storage Temperature:Ambient
  • UN:2929
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications