0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Cannulae
Disposable cannulas
Disposable cannulas
Disposable cannulas, G14, Green, 120 mm, +, 2×120 mm
EHRS15886511
:
EHRHARDT SOEHNE
Disposable cannulas
EHRS15886511
:
EHRHARDT SOEHNE
10+
Disposable cannulas
Disposable cannulas, G14, Green, 120 mm, +, 2×120 mm
130,00 €
1 * 100 items
-
+
Disposable cannulas.
Pyrogen-free, not toxic
With lancet cut
EO-sterilisation
Ø×L:
2×120 mm
Gauge:
G14
Sterile:
+
Colour:
Green
Description:
Disposable cannulas
Length:
120 mm
Seq:
30