Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
MODIFICATION KIT 100 ML SPRING 1 * 1 items
MODIFICATION KIT 100 ML SPRING 1 * 1 items
Print
MODIFICATION KIT 100 ML SPRING 1 * 1 items
Catalog # KNAUC40746V38
Supplier:
KNAUER
MODIFICATION KIT 100 ML SPRING 1 * 1 items
Catalog # KNAUC40746V38
Supplier:
KNAUER
Expand image
Return to Top
MODIFICATION KIT 100 ML SPRING 1 * 1 items
Add to cart
Price
598,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents