Orders LinkContactUsLinkComponent
Disposable cannulas
Disposable cannulas, G18, Pink, 100 mm, +, 1,2×100 mm
  EHRS15885691
Disposable cannulas
  EHRS15885691
Disposable cannulas
Disposable cannulas
Disposable cannulas, G18, Pink, 100 mm, +, 1,2×100 mm
114,00 €
1 * 100 items
Disposable cannulas.
  • Pyrogen-free, not toxic
  • With lancet cut
  • EO-sterilisation
  • Ø×L:
    1,2×100 mm
  • Gauge:
    G18
  • Sterile:
    +
  • Colour:
    Pink
  • Description:
    Disposable cannulas
  • Length:
    100 mm
  • Seq:
    22