Refrigerator, laboratory, convection cooling gross capacity 252l, power consumption 170w, lockable door energy consumption 1.4kWh, o uter dimension 121.6x60x60cm, inner dimension 104.7x51.3x44.1cm, with tc02 controller, indoor temperature range 0 de 1 * 1 items
Refrigerator, laboratory, convection cooling gross capacity 252l, power consumption 170w, lockable door energy consumption 1.4kWh, o uter dimension 121.6x60x60cm, inner dimension 104.7x51.3x44.1cm, with tc02 controller, indoor temperature range 0 de 1 * 1 items