Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bottles, narrow neck, with thread

1000 ml
  215-2384
 :   GRAVIS
Bottles, narrow neck, with thread
  215-2384
 :  GRAVIS
Bottles, narrow neck, with thread
Bottles, narrow neck, with thread

1000 ml

 
88,70 €
   
1 * 20 items
 

Glass, clear, amber or yellow.

  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances

Choice of various screw caps.

 : Supplied without screw caps - please order separately.

 : Packaged in impermeable plastic packs.

 
  • Capacity:
    1000 ml
  • Seq:
    10