Order Entry
Export
Orders LinkContactUsLinkComponent
Benzophenone imine 95%

Benzophenone imine 95%

Glass bottle for solids, 5 g
Catalog # H30110.06
Irritant
Benzophenone imine 95%
Catalog # H30110.06
CAS Number:  1013-88-3
Benzophenone imine 95%
Benzophenone imine 95%

Glass bottle for solids, 5 g

CAS Number:  1013-88-3
Price
84,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 95% CAS No.: 1013-88-3 Molecular Formula: C13H11N Formula Weight: 181.23 Refractive Index: 1.6180 Boiling Point: 151-153°/10mm MDL No.: MFCD00001760

Synonyms:  Benzophenoneimine
  • CAS Number:1013-88-3
  • Formula:C₁₃H₁₁N
  • MDL Number:MFCD00001760
  • Molecular Weight:181.24 g/mol
  • Boiling Pt:151…153 °C (10 mmHg)
  • Density:1.08
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications