Diagnostic vials, Nalgene®
20,00 ml, 30×62 mm, Yes, 20-415
Supplier: Thermo Fisher Scientific
Diagnostic vials, Nalgene®
Catalog # NALG342035-0020
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Includes lined, white HDPE closure for a leakproof seal
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached. These vials are bulk packed, 100 vials per tray, 500 vials per case.
Documents
Specifications
- Cap included:Yes
- Cap Size:20-415
- Ø×H:30×62 mm
- Capacity:20,00 ml
- Seq:03
- Cat. No.:NALG342035-0020
- Environmental certification:NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



