Orders LinkContactUsLinkComponent
Spouts for jerricans
Spouts for jerricans
Rigid spout, electroconductive, S60/61
Catalog # SCTV610504
Supplier:   S.C.A.T.
Spouts for jerricans
Catalog # SCTV610504
Supplier:  S.C.A.T.
Spouts for jerricans
Spouts for jerricans
Rigid spout, electroconductive, S60/61
Price
201,99 €
Log in for availability
1 * 1 items
Product Details & Documents
For simple and safe pouring of fluids from containers.
  • Plastic
  • Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Specifications
  • Thread:
    S60/61
  • Description:
    Rigid spout, electroconductive
  • Seq:
    05