Order Entry
Export
Orders LinkContactUsLinkComponent
Politainer

Politainer

Politainer, 20 L, Natural, S60/61, UN-Y approval in combination with box, 285×285×285 mm
Catalog # SCTV107333
Supplier:   S.C.A.T.
Politainer
Catalog # SCTV107333
Supplier:  S.C.A.T.
Politainer
Politainer

Politainer, 20 L, Natural, S60/61, UN-Y approval in combination with box, 285×285×285 mm

Price
15,12 €
Log in for availability
1 * 1 items
Product Details & Documents

The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.

  • Consists of LDPE and EVA copolymer, which offers optimum protection against volatile solvents
  • Available with the frequently used threads GL38 and S60/61 and with volumes of 5, 10 and 20 L

SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.

 : The matching boxes with UN-Y approval are available in the table below.

Specifications
  • Thread:
    S60/61
  • Type:
    UN-Y approval in combination with box
  • Capacity:
    20 L
  • Colour:
    Natural
  • Description:
    Politainer
  • Seq:
    04
  • W×D×H:
    285×285×285 mm
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Thread adapter, PTFE, GL 38 on GL 45
    Thread adapter, PTFE, GL 38 on GL 45
    SCTV107338
    Foldable politainer with GL 38
    Log in for availability
    1 * 1 items - 65,87 €