Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Thermal hl40 bath fluid 5 liters (-40...+250 c) 1 * 5 l
Catalog # 461-0480
Supplier:
JULABO GmbH
Expand image
Thermal hl40 bath fluid 5 liters (-40...+250 c) 1 * 5 l
Catalog # 461-0480
Supplier:
JULABO GmbH