Orders LinkContactUsLinkComponent
Printer and balance accessories
Printer and balance accessories
Printer cable (SF40A) for the CW11, CKW55 and TxxW indicators
Catalog # 611-2574
Supplier:   OHAUS
Printer and balance accessories
Catalog # 611-2574
Supplier:  OHAUS
Printer and balance accessories
Printer and balance accessories
Printer cable (SF40A) for the CW11, CKW55 and TxxW indicators
Price
102,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The SF40A impact printer and the STP103 thermal printer are compatible with any Ohaus balance with an RS232 interface. They are particularly useful when coupled to an Ohaus balance with GLP functionality as they can provide a hard copy audit trail. A printer cable is required (but not supplied) for specific Ohaus balances.
Ordering information: There are a wide variety of accessories and replacement parts for Ohaus balances and the most commonly used are listed below: For further items please visit the local Avantor website.
Specifications
  • Description:
    Printer cable (SF40A) for the CW11, CKW55 and TxxW indicators
  • Seq:
    20