ContactUsLinkComponent
Microplate 384 well PCR white with sealing film barcode non-sterile compatible with Roche Light Cycler 480 1 * 10 items
Catalog # 732-1220
Supplier:  Corning
Microplate 384 well PCR white with sealing film barcode non-sterile compatible with Roche Light Cycler 480 1 * 10 items
Catalog # 732-1220
Supplier:  Corning