Volumetric flasks, class A, white markings
25* ml, 12/21, 0,060 ± ml
612-4715
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
- White enamel indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
: DIN EN ISO 1042
- NS:12/21
- Capacity:25* ml
- Tolerance:0,060 ± ml